missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRGAP2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112857
This item is not returnable.
View return policy
Description
SRGAP2B Polyclonal Antibody for Western Blot, ICC/IF, ELISA
This locus encodes a member of the SLIT-ROBO Rho GTPase activating protein family. This human-specific locus resulted from incomplete segmental duplication of the SLIT-ROBO Rho GTPase activating protein 2 locus. The encoded protein lacks the GTPase activating protein domain compared to proteins encoded by SLIT-ROBO Rho GTPase activating protein 2. The functionality of the protein encoded by this locus has been questioned, as several normal individuals with homozygous deletions for this locus have been identified, and the expression of this locus appears to be much lower than the similar SLIT-ROBO Rho GTPase activating protein 2C (SRGAP2C) locus. The SRGAP2C locus has been shown to encode a protein that functions antagonistically to SLIT-ROBO Rho GTPase activating protein 2 in cortical neuron development. [provided by RefSeq, Dec 2012].
Specifications
| SRGAP2B | |
| Polyclonal | |
| Unconjugated | |
| SRGAP2B | |
| SLIT-ROBO Rho GTPase activating protein 2 pseudogene 2; SLIT-ROBO Rho GTPase activating protein 2B; SLIT-ROBO Rho GTPase activating protein 2B (pseudogene); SLIT-ROBO Rho GTPase-activating protein 2B; SRGAP2B; SRGAP2L; SRGAP2P2 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 647135 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P0DMP2 | |
| SRGAP2B | |
| Recombinant Human SLIT-ROBO Rho GTPase-activating protein 2B protein (79-150AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction