Learn More
Invitrogen™ SRP19 Recombinant Rabbit Monoclonal Antibody (24GB1815)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557249
 View more versions of this product
Product Code. 30276055
  • € 500.00 / 100µL
Description
SRP19 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
SRP19 belongs to the SRP19 family. It is signal-recognition-particle assembly and binds directly to 7S RNA and mediates binding of the 54 kDa subunit of the SRP.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SRP19
Recombinant Monoclonal
Unconjugated
SRP19
2310020D23Rik; hypothetical protein LOC514960; signal recognition particle; signal recognition particle 19; signal recognition particle 19 kDa protein; signal recognition particle 19kD; signal recognition particle 19kDa; SRP19; zgc:63961
Rabbit
Affinity Chromatography
RUO
291685, 66384, 6728
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1815
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P09132, Q9D7A6
SRP19
A synthesized peptide derived from human SRP19 (aa 10-46).
100 μL
Primary
Human, Mouse, Rat