missing translation for 'onlineSavingsMsg'
Learn More

SRp20 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16827623 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16827623 20 μL 20µL
16817623 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16827623 Supplier Proteintech Supplier No. 109161AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SR proteins appear to be recruited from nuclear "speckles", in which they are concentrated, to sites of transcription in order to spatially coordinate transcription and pre-mRNA splicing within the cell nucleus.

Specifications

Antigen SRp20
Applications Immunohistochemistry, ELISA
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SRSF3
Gene Accession No. P84103, P84104
Gene Alias Pre mRNA splicing factor SRP20, SFRS3, SRp20, SRSF3
Gene Symbols Srsf3
Host Species Rabbit
Immunogen SRp20 Fusion Protein Ag1341
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20383, 361814, 6428
Target Species Mouse, Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.