missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRP54 Antibody (SR1965), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22000-100ul
This item is not returnable.
View return policy
Description
SRP54 Monoclonal antibody specifically detects SRP54 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Specifications
| SRP54 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
| SR1965 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
| signal recognition particle 54 kDa protein, signal recognition particle 54kD, signal recognition particle 54kDa | |
| A synthesized peptide derived from human SRP54 (Uniprot #: P61011) | |
| 100 μg | |
| Signal Transduction | |
| 6729 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction