missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SRP54 Recombinant Rabbit Monoclonal Antibody (1O3J1)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA542773

Product Code. 17838092

  • € 528.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive test controls include: HeLa, Jurkat. Immunogen sequence: PGRIQRVARG SGVSTRDVQE LLTQYTKFAQ MVKKMGGIKG LFKGGDMSKN VSQSQMAKLN QQMAKMMDPR VLHHMGGMAG LQSMMRQFQQ GAAGNMKGMM GFNNM.

Binds to the signal sequence of presecretory protein when they emerge from the ribosomes and transfers them to TRAM (translocating chain-associating membrane protein).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SRP54
Recombinant Monoclonal
0.4 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P61011, Q6AYB5
SRP54, Srp54a
A synthetic peptide corresponding to a sequence within amino acids 400-504 of human SRP54 (P61011).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Western Blot, Immunocytochemistry
1O3J1
Unconjugated
SRP54
54K subunit of signal recognition particle; 54kDa; signal recognition particle 54; signal recognition particle 54 kDa protein; signal recognition particle 54A; signal recognition particle 54kD; signal recognition particle 54kDa; SRP54; Srp54a
Rabbit
Affinity chromatography
RUO
116650, 24067, 6729
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.