missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRp55 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-04142IR
This item is not returnable.
View return policy
Description
SRp55 Polyclonal antibody specifically detects SRp55 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SRp55 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| B52, FLJ08061, pre-mRNA splicing factor SRP55, Pre-mRNA-splicing factor SRP55, serine/arginine-rich splicing factor 6, SFRS6, Splicing factor, arginine/serine-rich 6MGC5045, splicing factor, arginine/serine-rich, 55 kDa, SR splicing factor 6, SRP55arginine/serine-rich splicing factor 6 | |
| The immunogen recognized by this antibody maps to this antibody using the numbering given in entry NP_006266.2 (GeneID 6431). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 6431 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction