missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRP68 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22307
This item is not returnable.
View return policy
Description
SRP68 Polyclonal specifically detects SRP68 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| SRP68 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| signal recognition particle 68 kDa protein, signal recognition particle 68kD, signal recognition particle 68kDa | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 6730 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg | |
| NP_055045.2 | |
| SRP68 | |
| The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Signal Recognition Particle 68kDa using the numbering given in entry NP_055045.2 (GeneID 6730). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction