missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRP9 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-61107AF700
This item is not returnable.
View return policy
Description
SRP9 Polyclonal antibody specifically detects SRP9 in Human samples. It is validated for Western Blot
Specifications
| SRP9 | |
| Polyclonal | |
| Western Blot | |
| ALURBP, MGC110981, signal recognition particle 9 kDa protein, signal recognition particle 9kD, signal recognition particle 9kDa | |
| A region between residue 36 to 86 of human Signal recognition particle 9 kDa protein. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6726 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction