missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRPK2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97313-100ul
This item is not returnable.
View return policy
Description
SRPK2 Polyclonal antibody specifically detects SRPK2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SRPK2 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| EC 2.7.11.1, FLJ36101, H_RG152G17.1a, H_RG152G17.1b, serine kinase SRPK2, Serine/arginine-rich protein-specific kinase 2, serine/arginine-rich splicing factor kinase 2, serine/threonine-protein kinase SRPK2, SFRS protein kinase 2SFRSK2, SR protein kinase 2, SR-protein-specific kinase 2, SRSF protein kinase 2, WUGSC:H_RG152G17.1a | |
| Produced in rabbits immunized with E. coli-derived Human SRPK2 fragment. | |
| 100 μg | |
| Protein Kinase | |
| 6733 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction