missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SRPR alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-28900
This item is not returnable.
View return policy
Description
SRPR alpha Polyclonal antibody specifically detects SRPR alpha in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| SRPR alpha | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Docking protein alpha, DP, DP-alpha, MGC17355, MGC3650, MGC9571, signal recognition particle receptor (docking protein), signal recognition particle receptor ('docking protein'), signal recognition particle receptor subunit alpha, Sralpha, SR-alpha, SRP-alpha | |
| Between 225 and 275 | |
| 100 μg | |
| Signal Transduction | |
| 6734 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction