missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SRPRB Polyclonal Antibody
GREENER_CHOICE

Product Code. 15750751 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15750751 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15750751 Supplier Invitrogen™ Supplier No. PA552802

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LCDSGKTLLF VRLLTGLYRD TQTSITDSCA VYRVNNNRGN SLTLIDLPGH ESLRLQFLER FKSSARAIVF VVDSAAFQRE VKDVAEFLYQ VLIDSMGLKN TPSFLIACNK QDIAMAKSAK LIQQQLEKEL NTL Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

SRPRB has similarity to mouse protein which is a subunit of the signal recognition particle receptor (SR). This subunit is a transmembrane GTPase belonging to the GTPase superfamily. It anchors alpha subunit, a peripheral membrane GTPase, to the ER membrane. SR is required for the cotranslational targeting of both secretory and membrane proteins to the ER membrane. The protein encoded by this gene has similarity to mouse protein which is a subunit of the signal recognition particle receptor (SR). This subunit is a transmembrane GTPase belonging to the GTPase superfamily. It anchors alpha subunit, a peripheral membrane GTPase, to the ER membrane. SR is required for the cotranslational targeting of both secretory and membrane proteins to the ER membrane. The protein encoded by this gene has similarity to mouse protein which is a subunit of the signal recognition particle receptor (SR). This subunit is a transmembrane GTPase belonging to the GTPase superfamily. It anchors alpha subunit, a peripheral membrane GTPase, to the ER membrane. SR is required for the cotranslational targeting of both secretory and membrane proteins to the ER membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRPRB
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SRPRB
Gene Accession No. Q9Y5M8
Gene Alias AA409543; Ab2-417; APMCF1; Ba1-667; Cc1-8; hypothetical protein; Protein APMCF1; PSEC0230; Signal recognition particle receptor subunit beta; signal recognition particle receptor subunit beta pseudogene; signal recognition particle receptor, B subunit; signal recognition particle receptor, beta subunit; SR-beta; SRP receptor beta subunit; SRP receptor subunit beta; SRPRB; wu:fk57h10; zgc:111820; zgc:92746; zgc:92746 protein
Gene Symbols SRPRB
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SRPRB. Recombinant protein control fragment (Product #RP-90317).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58477
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.