missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRRM3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553971
This item is not returnable.
View return policy
Description
Immunogen sequence: QGYSEEEIRQ KVGTFRQMLM EKEGVLTRED RPGGHIVAET PRLTEGAEPG LEYAPFDDDD GPVDCDCPAS CYRGHRGYRT KH Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 88%.
Specifications
| SRRM3 | |
| Polyclonal | |
| Unconjugated | |
| SRRM3 | |
| 2900083I11Rik; AB041661; MNCb-4760; RGD1307391; serine/arginine repetitive matrix 3; serine/arginine repetitive matrix protein 3; SRm300-like; Srrm2l; SRRM2-like protein; Srrm3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 222183 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A6NNA2 | |
| SRRM3 | |
| Recombinant protein corresponding to Human SRRM3. Recombinant protein control fragment (Product #RP-92847). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction