missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRRM4 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06263APC
This item is not returnable.
View return policy
Description
SRRM4 Polyclonal antibody specifically detects SRRM4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SRRM4 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| KIAA1853, medulloblastoma antigen MU-MB-2.76, MU-MB-2.76, neural-specific SR-related protein of 100 kDa, nSR100, serine/arginine repetitive matrix 4, serine/arginine repetitive matrix protein 4 | |
| Produced in rabbits immunized with E. coli-derived Human SRRM4 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 84530 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction