missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRrp35 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569114
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 85%; Rabbit: 100%; Rat: 100%.
SRSF12 gene ontology annotations related to this gene include regulation of alternative mRNA splicing, via spliceosome.
Specifications
| SRrp35 | |
| Polyclonal | |
| Unconjugated | |
| SRSF12 | |
| 35 kDa serine-arginine repressor; 35 kDa SR repressor protein; 35 kDa SR repressor protein-like protein; B830039L16; RGD1561162; RP11-63L7.3; serine and arginine rich splicing factor 12; serine/arginine-rich splicing factor 12; serine/arginine-rich splicing factor 13B; serine-arginine repressor protein; serine-arginine repressor protein (35 kDa); serine-arginine repressor protein 35; Sfrs13b; SFRS19; splicing factor, arginine/serine-rich 13B; Splicing factor, arginine/serine-rich 19; SR splicing factor 12; Srrp; SRRP35; Srsf12; Srsf13b | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 135295 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8WXF0 | |
| SRSF12 | |
| synthetic peptide directed towards the middle region of human SRrp35. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction