missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SRSF3 Recombinant Rabbit Monoclonal Antibody (K01_2Q45)

Product Code. 30828917
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828917 50 μL
30826263 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828917 Supplier Invitrogen™ Supplier No. MA565728

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: EFEDPRDAAD AVRE.

SR proteins appear to be recruited from nuclear 'speckles', in which they are concentrated, to sites of transcription in order to spatially coordinate transcription and pre-mRNA splicing within the cell nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRSF3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_2Q45
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene SRSF3
Gene Accession No. P84103, P84104
Gene Alias AL024116; pre-mRNA splicing factor SRp20; pre-mRNA-splicing factor SRP20; Protein X16; serine and arginine rich splicing factor 3; serine/arginine-rich splicing factor 3; SFRS3; splicing factor, arginine/serine-rich 3; splicing factor, arginine/serine-rich 3 (SRp20); splicing factor, arginine/serine-rich, 20-kD; Srp20; SRSF3; X16
Gene Symbols SRSF3
Host Species Rabbit
Immunogen A synthetic peptide of human SFRS3 (aa 52-65).
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20383, 361814, 6428
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.