missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRSF9 Mouse anti-Human, Clone: OTI5G7, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808424
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the serine/arginine (SR)-rich family of pre-mRNA splicing factors, which constitute part of the spliceosome. Each of these factors contains an RNA recognition motif (RRM) for binding RNA and an RS domain for binding other proteins. The RS domain is rich in serine and arginine residues and facilitates interaction between different SR splicing factors. In addition to being critical for mRNA splicing, the SR proteins have also been shown to be involved in mRNA export from the nucleus and in translation. Two pseudogenes, one on chromosome 15 and the other on chromosome 21, have been found for this gene.Specifications
| SRSF9 | |
| Monoclonal | |
| Unconjugated | |
| SRSF9 | |
| SFRS9, SRp30c | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8683 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5G7 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13242 | |
| SRSF9 | |
| Full length human recombit protein of human SRSF9 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction