Learn More
SRX1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14273-1-AP-150UL
 View more versions of this product
Product Code. 16842844
  • € 410.00 / 150µL
Specifications
SRX1
Polyclonal
Unconjugated
SRXN1
C20orf139, SRX, SRX1, SRXN1, Sulfiredoxin 1, YKL086W
Rabbit
Antigen Affinity Chromatography
RUO
140809, 76650
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BYN0, Q9D975
Srxn1
SRX1 Fusion Protein Ag5613
150 μL
Primary
Human, Mouse