missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRY Antibody (OTI3C8), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74352F
This item is not returnable.
View return policy
Description
SRY Monoclonal specifically detects SRY in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| SRY | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| SRY | |
| Full length recombinant protein of human SRY (NP_003131) produced in E.coli. | |
| 0.1 mL | |
| 6736.0 | |
| Store at 4°C in the dark. |
| Immunocytochemistry, Immunofluorescence | |
| OTI3C8 | |
| Immunocytochemistry/Immunofluorescence | |
| essential protein for sex determination in human males, sex determining region Y, sex-determining region on Y, sex-determining region Y protein, TDFsex determining region protein, TDY, Testis-determining factor | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction