missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SS18 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98476R
This item is not returnable.
View return policy
Description
SS18 Polyclonal specifically detects SS18 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| SS18 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| MGC116875, Protein SYT, SSXTsynovial sarcoma, translocated to X chromosome, Synovial sarcoma translocated to X chromosome protein, synovial sarcoma translocation, chromosome 18, SYTprotein SSXT | |
| Produced in rabbits immunized with E. coli-derived Human SS18 fragment. | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 6760 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction