missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SS18L2 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98477IR
This item is not returnable.
View return policy
Description
SS18L2 Polyclonal antibody specifically detects SS18L2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SS18L2 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| KIAA-iso, SS18-like protein 2, synovial sarcoma translocation gene on chromosome 18-like 2, SYT homolog 2 | |
| Produced in rabbits immunized with E. coli-derived Human SS18L2 fragment. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 51188 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction