missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSB Antibody (OTI2C8), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74356B
This item is not returnable.
View return policy
Description
SSB Monoclonal specifically detects SSB in Human, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| SSB | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| SSB | |
| Full-length protein expressed in 293T cell transfected with human SSB expression vector. | |
| 0.1 mL | |
| Primary | |
| Human, Canine, Primate, Monkey | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI2C8 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| La autoantigen, La ribonucleoprotein, lupus La protein, member 3, Sjoegren syndrome type B antigen, Sjogren syndrome antigen B (autoantigen La), SS-B, SS-B/La protein | |
| Mouse | |
| Affinity Purified | |
| Core ESC Like Genes, Stem Cell Markers | |
| 6741.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction