missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSB Mouse anti-Canine, Human, Clone: OTI3H9, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500410
This item is not returnable.
View return policy
Description
La is involved in diverse aspects of RNA metabolism, including binding and protecting 3-prime UUU(OH) elements of newly RNA polymerase III-transcribed RNA, processing 5-prime and 3-prime ends of pre-tRNA precursors, acting as an RNA chaperone, and binding viral RNAs associated with hepatitis C virus. La protein was originally defined by its reactivity with autoantibodies from patients with Sjogren syndrome and systemic lupus erythematosus.Specifications
| SSB | |
| Monoclonal | |
| 1.2 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P05455 | |
| SSB | |
| Full-length protein expressed in 293T cell transfected with human SSB expression vector | |
| 100 μL | |
| Primary | |
| Canine, Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| OTI3H9 | |
| Unconjugated | |
| SSB | |
| LARP3, La, La/SSB | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 478787, 6741 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction