missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12212-1-AP-150UL
This item is not returnable.
View return policy
Description
This protein binds preferentially and cooperatively to ss-DNA. Probably involved in mitochondrial DNA replication. SSBP is a housekeeping gene involved in mitochondrial biogenesis (Tiranti et al. 1995).Specifications
| SSBP1 | |
| Polyclonal | |
| Unconjugated | |
| SSBP1 | |
| Mt SSB, MtSSB, PWP1 interacting protein 17, SSBP, SSBP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 381760, 54304, 6742 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P28042, Q04837, Q9CYR0 | |
| Ssbp1 | |
| SSBP1 Fusion Protein Ag2860 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction