missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSBP3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 483.00
Specifications
| Antigen | SSBP3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
SSBP3 Polyclonal specifically detects SSBP3 in Human samples. It is validated for Western Blot.Specifications
| SSBP3 | |
| Polyclonal | |
| Rabbit | |
| Q5T860 | |
| 23648 | |
| Synthetic peptides corresponding to SSBP3(single stranded DNA binding protein 3) The peptide sequence was selected from the middle region of SSBP3. Peptide sequence SNFPMGPGSDGPMGGMGGMEPHHMNGSLGSGDIDGLPKNSPNNISGISNP. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| CSDP, FLJ10355, single stranded DNA binding protein 3, SSDP1Sequence-specific single-stranded-DNA-binding protein, SSDPsingle-stranded DNA-binding protein 3 | |
| SSBP3 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title