missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSC5D Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-93831
This item is not returnable.
View return policy
Description
SSC5D Polyclonal specifically detects SSC5D in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SSC5D | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| scavenger receptor cysteine rich domain containing (5 domains), scavenger receptor cysteine-rich glycoprotein, soluble scavenger protein with 5 SRCR domains, soluble scavenger receptor cysteine-rich domain-containing protein SSC5D, soluble scavenger with 5 domains | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 284297 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| SSC5D | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:TCTGYTDYDDYPPWTWDPTSREDLAKGTTTAGVPGHTLPWRTTRRPGSSSPAIRRLPDTGSKDGYKLPWTWDTPSGRGLAEGTPTAGKLGPTL | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto