missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSEA-1 Antibody (FUT4/1478R), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 353.00 - € 587.00
Specifications
| Antigen | SSEA-1 |
|---|---|
| Clone | FUT4/1478R |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1-2 μg/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL, Immunofluorescence 0.5-1ug/mL |
| Applications | Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18629046
|
Novus Biologicals
NBP2-49878-0.1mg |
0.1 mg |
€ 587.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18647247
|
Novus Biologicals
NBP2-49878SS |
0.02 mg |
€ 353.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SSEA-1 Monoclonal antibody specifically detects SSEA-1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), ImmunofluorescenceSpecifications
| SSEA-1 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Pancreatic Stem Cell Markers, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| alpha-(1,3)-fucosyltransferase, CD15, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.65, ELAM-1 ligand fucosyltransferase, ELFT, FCT3AELAM ligand fucosyltransferase, Fucosyltransferase 4, fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific), Fucosyltransferase IV, fucT-IV, FUC-TIV, FUTIV, Galactoside 3-L-fucosyltransferase, Lewis X, LeX, SSEA-1, stage-specific embryonic antigen 1 | |
| Recombinant human full-length SSEA-1 protein (Uniprot: P22083) | |
| Primary | |
| Store at 4°C. |
| FUT4/1478R | |
| Flow Cytometry 1-2 μg/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL, Immunofluorescence 0.5-1ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| P22083 | |
| 2526 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title