missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSEA-3 Antibody (MC-631) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-80962
This item is not returnable.
View return policy
Beschreibung
SSEA-3 Monoclonal specifically detects SSEA-3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.
Spezifikation
| SSEA-3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500, Flow Cytometry : 10-20 μg/mL, Immunohistochemistry : 1:10-1:500, Immunocytochemistry/Immunofluorescence : 1:50-1:100, Immunoprecipitation : 1:10-1:500, Immunohistochemistry-Paraffin : 1:10-1:500, Immunohistochemistry-Frozen : 1:10-1:500, CyTOF-ready | |
| Stage specific embryonic antigen 3, Stage-specific embryonic antigen 3 | |
| 4-8 cell stage mouse embryos. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| MC-631 | |
| Unconjugated | |
| PBS | |
| Rat | |
| IgM purified | |
| RUO | |
| This antibody recognizes the SSEA3 that is expressed upon the surface of human teratocarcinoma stem cells (EC), human embryonic germ cells (EG) and human embryonic stem cells (ES). No immunoreactivity is evident with undifferentiated murine EC, ES and EG cells. Expression of SSEA3 is down regulated following differentiation of human EC cells. In contrast, the differentiation of murine EC and ES cells may be accompanied by an increase in SSEA-3 expression. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgM |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur