missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSEA-4 Antibody (MC-813-70), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-26644AF488
This item is not returnable.
View return policy
Description
SSEA-4 Monoclonal specifically detects SSEA-4 in Human, Mouse, Canine, Chicken, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface).
Specifications
| SSEA-4 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| NP_803131 | |
| Human embryonal carcinoma cell line 2102Ep | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Canine, Chicken, Primate, Rabbit | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| MC-813-70 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface) | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Chicken, Dog, Human, Mouse, New World Monkey, Rabbit. | |
| Store at 4C in the dark. | |
| IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction