Learn More
SSEA4 Monoclonal Antibody (eBioMC-813-70 (MC-813-70)), eFluor™ 570, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 41-8843-80
Additional Details : Weight : 0.25000kg
Description
Description: The eBioMC-813-70 monoclonal antibody reacts with a glycolipid carbohydrate epitope on Stage-specific embryonic antigen-4 (SSEA-4) protein. Expression is found on human teratocarcinoma stem cells (EC), human embryonic germ cells (EG), human induced pluripotent stem (iPS) cells, and human embryonic stem cells (ES) with a decrease in expression upon differentiation. Recently expression has also been found on bone marrow mesenchymal stem cells. The eBioMC-813-70 antibody has been reported to crossreact to mouse, rabbit, canine, chicken, and Old and New World monkeys. Applications Reported: This eBioMC-813-70 (MC-813-70) antibody has been reported for use in immunocytochemistry. This product has not been validated for flow cytometric analysis. Applications Tested: This eBioMC-813-70 (MC-813-70) antibody has been tested by immunocytochemistry on fixed NTERA-2 and 2102Ep cells and can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filter Recommendation: When using this eFluor™ 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor™ 555 or TRITC filter is acceptable. Excitation: 555 nm; Emission: 570 nm
Description: The eBioMC-813-70 monoclonal antibody reacts with a glycolipid carbohydrate epitope on Stage-specific embryonic antigen-4 (SSEA-4) protein. Expression is found on human teratocarcinoma stem cells (EC), human embryonic germ cells (EG), human induced pluripotent stem (iPS) cells, and human embryonic stem cells (ES) with a decrease in expression upon differentiation. Recently expression has also been found on bone marrow mesenchymal stem cells.The eBioMC-813-70 antibody has been reported to crossreact to mouse, rabbit, canine, chicken, and Old and New World monkeys.Applications Reported: This eBioMC-813-70 (MC-813-70) antibody has been reported for use in immunocytochemistry. This product has not been validated for flow cytometric analysis.Applications Tested: This eBioMC-813-70 (MC-813-70) antibody has been tested by immunocytochemistry on fixed NTERA-2 and 2102Ep cells and can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filter Recommendation: When using this eFluor™ 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor™ 555 or TRITC filter is acceptable.Excitation: 555 nm; Emission: 570 nmSpecifications
SSEA4 | |
Monoclonal | |
0.2mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
Mouse | |
25 μg | |
Primary | |
Store at 2-8°C. Do not freeze. Light-sensitive material. | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
eBioMC-813-70 (MC-813-70) | |
eFluor 570 | |
stage-specific embryonic antigen 4, SSEA-4 | |
Affinity chromatography | |
RUO | |
Canine, Chicken, Human, Mouse, Non-human Primate, Rabbit | |
Antibody | |
IgG3 |
For Research Use Only.