Learn More
Invitrogen™ SSEA4 Monoclonal Antibody (MC-813-70)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA1021X
This item is not returnable.
View return policy
Description
SSEA4 Monoclonal Antibody for Flow, ICC/IF
SSEA-4 (Stage-specific embryonic antigen 4) is a cell surface antigen, expressed along with SSEA-3, TRA-1-60 and TRA-1-81 in embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). In human cells, these surface markers are down-regulated during the differentiation process, while SSEA-1 is absent in undifferentiated human stem cells and upregulated on the cell surface after retinoic acid mediated differentiation. Conversely, in mouse stem cells, SSEA-1 expression decreases with differentiation while SSEA-3 and SSEA-4 expression increase.
Specifications
| SSEA4 | |
| Monoclonal | |
| 1 mg/mL | |
| Potassium phosphate with 0.15M NaCl, 1mg/mL BSA and 0.05% sodium azide | |
| Mouse | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG3 |
| Flow Cytometry, Immunocytochemistry | |
| MC-813-70 | |
| Unconjugated | |
| SSEA-4; stage-specific embryonic antigen 4 | |
| human embryonal carcinoma cell line 2102Ep. | |
| 20 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |