missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSFA2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-88367
This item is not returnable.
View return policy
Description
SSFA2 Polyclonal specifically detects SSFA2 in Human samples. It is validated for Western Blot.
Specifications
| SSFA2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| ITPRID2 | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of human SSFA2 | |
| 100 μL | |
| Cell Cycle and Replication | |
| 6744 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Cleavage signal-1 protein, CS1, CS-1DKFZp313O1039, FLJ45996, KIAA1927, Ki-ras-induced actin-interacting protein, KRAPDKFZp779G0129, SPAG13, sperm specific antigen 2, sperm-specific antigen 2 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction