Learn More
Invitrogen™ SSFA2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110432
Description
Immunogen sequence: EVDQLQGLRN SVRMELQDLE LQLEERLLGL EEQLRAVRMP SPFRSSALMG MCGSRSADNL SCPSPLNVME PVTELMQEQS YLKSELGLGL GEMGFEIPPG ESSESVFSQA TSESSSVCSG PSHANRRTGV PSTASVGKSK TPLVARKKVF RASVALTPTA PSRTGSVQTP PDLESSEEVD AAEGAPEVVG PKSEVEEGHG KLPSMPAAEE MHKNVEQDEL QQVIREIKES IVGEIRREIV SGLLAAVSSS KASNSKQDYH.
SSFA2 gene ontology annotations related to this gene include actin binding.
Specifications
| SSFA2 | |
| Polyclonal | |
| Unconjugated | |
| ITPRID2 | |
| 5730488C15Rik; cleavage signal-1 protein; CS1; CS-1; ITPRID2; ITPR-interacting domain-containing protein 2; KIAA1927; Ki-ras-induced actin-interacting protein; Krap; KRAS-induced actin-interacting protein; mKIAA1927; Protein ITPRID2; SPAG13; sperm associated antigen 13; sperm specific antigen 2; sperm-specific antigen 2; Sperm-specific antigen 2 homolog; SSFA2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 70599 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.83 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q922B9 | |
| ITPRID2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1000-1259 of human SSFA2 (NP_001123917.1). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.