Learn More
SSPN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26685-1-AP-20UL
 View more versions of this product
Product Code. 16825085
  • € 145.00 / 20µL
Description
Component of the dystrophin-glycoprotein complex (DGC), a complex that spans the muscle plasma membrane and forms a link between the F-actin cytoskeleton and the extracellular matrix. Preferentially associates with the sarcoglycan subcomplex of the DGC.
Specifications
Specifications
SSPN
Polyclonal
Unconjugated
SSPN
DAGA5, Kirsten ras associated protein, KRAG, ospan, NSPN, Sarcospan, SPN1, SPN2, SSPN
Rabbit
Antigen Affinity Chromatography
RUO
16651, 500364, 8082
-20°C
Liquid
Western Blot
0.67 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q14714, Q62147
Sspn
SSPN Fusion Protein Ag24511
20 μL
Primary
Human, Mouse, Rat