missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSRP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15696-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a protein that belongs to the microtubule-associated protein family. The proteins of this family are thought to be involved in microtubule assembly, which is an essential step in neurogenesis. The product of this gene is a precursor polypeptide that presumably undergoes proteolytic processing to generate the final MAP1B heavy chain and LC1 light chain. Gene knockout studies of the mouse microtubule-associated protein 1B gene suggested an important role in development and function of the nervous system.Specifications
| SSRP1 | |
| Polyclonal | |
| Unconjugated | |
| SSRP1 | |
| FACT, FACT 80 kDa subunit, FACT complex subunit SSRP1, FACT80, FACTp80, hSSRP1, SSRP1, T160 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20833, 6749, 81785 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q04931, Q08943, Q08945 | |
| SSRP1 | |
| SSRP1 Fusion Protein Ag8318 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction