Learn More
Invitrogen™ SSTR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120328
Description
Positive test controls include: mouse kidney, mouse liver. The target is usually found in the following locations: Cell membrane, Cytoplasm, Multi-pass membrane protein. Immunogen sequence: LPFYIFNVSS VSMAISPTPA LKGMFDFVVV LTYANSCANP ILYAFLSDNF KKSFQNVLCL VKVSGTDDGE RSDSKQDKSR LNETTETQRT LLNGDLQTSI.
SSTR2 (somatostatin receptor 2) is a member of the superfamily of receptors having seven transmembrane segments and is expressed in highest levels in the cerebrum and kidney. Somatostatin acts at many sites to inhibit the release of many hormones and other secretory proteins. The receptor is coupled via pertussis toxin sensitive G proteins to inhibit adenylyl cyclase.
Specifications
| SSTR2 | |
| Polyclonal | |
| Unconjugated | |
| SSTR2 | |
| Smstr2; Smstr-2; somatostatin receptor; somatostatin receptor 2; somatostatin receptor 2a; somatostatin receptor subtype 2; somatostatin receptor type 2; somatotropin release-inhibiting factor receptor; SRIF-1; SS2R; SS-2-R; SS2-R; SSR2; sst2; Sstr2; SSTR-2; SSTR2A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 20606, 54305, 6752 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.17 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P30680, P30874, P30875 | |
| SSTR2 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.