missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSTR5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 51029-2-AP-150UL
This item is not returnable.
View return policy
Description
Somatostatin is a multifunctional protein affecting exocrine and endocrine protein secretion, neurotransmission, and neuronal modulation and regulating cell proliferation. All these functions of Somatostatin are executed through a class of GPCR protein family members called somatostatin receptor. Till date, 6 classes of somatostatin receptors have been discovered out of which SSTR5, 364A.A protein, is a receptor subtype expressed predomitly in the pituitary and hypothalamus region of brain. Studies revealed that SSTR5 contains seven transmembrane regions and undergoes homodimerization and also heterodimerizes with SSTR1. Although the specific functions of SSTR5 are still elusive, the studies indicate that it complements SSTR2 regulation of ACTH secretion. Polymorphisms in SSTR5 gene have been observed in patients with acromegaly.Specifications
| SSTR5 | |
| Polyclonal | |
| Unconjugated | |
| SSTR5 | |
| somatostatin receptor 5, Somatostatin receptor type 5, SS 5 R, SS5 R, SS5R, SSTR5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20609, 6755 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O08858, P35346 | |
| SSTR5 | |
| SSTR5 Fusion Protein Ag0519 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction