missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ssu72 Antibody (45E2), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP2-60233IR
This item is not returnable.
View return policy
Description
Ssu72 Monoclonal antibody specifically detects Ssu72 in Human samples. It is validated for Western Blot, ELISA
Specifications
| Ssu72 | |
| Monoclonal | |
| DyLight 755 | |
| CTD phosphatase SSU72, EC 3.1.3.16, FLJ13048, HSPC182, PNAS-120, RNA polymerase II subunit A C-terminal domain phosphatase SSU72, SSU72 RNA polymerase II CTD phosphatase homolog (S. cerevisiae), Ssu72 RNA polymerase II CTD phosphatase homolog (yeast) | |
| Recombinant human SSU72 (1-194aa) purified from E. coli | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing, Proteases & Other Enzymes | |
| 29101 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA | |
| 4500 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction