missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSU72 Monoclonal Antibody (OTI10A4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809163S
This item is not returnable.
View return policy
Description
SSU72 Monoclonal antibody specifically detects SSU72 in Human samples. It is validated for Western BlotSpecifications
| SSU72 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NP77 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 29101 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI10A4 | |
| Unconjugated | |
| Ssu72 | |
| 1190002E22Rik; 1500011L16Rik; 2610101M12Rik; CTD phosphatase SSU72; FLJ13048; HSPC182; PNAS-120; RGD1307854; RNA polymerase II subunit A C-terminal domain phosphatase SSU72; RP5-832C2.2; SSU72; SSU72 homolog, RNA polymerase II CTD phosphatase; SSU72 RNA polymerase II CTD phosphatase homolog (S. cerevisiae); Ssu72 RNA polymerase II CTD phosphatase homolog (yeast) | |
| Full length human recombinant protein of human SSU72 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction