missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSX1 Antibody (OTI1E10), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74362MFV610
This item is not returnable.
View return policy
Description
SSX1 Monoclonal antibody specifically detects SSX1 in Human samples. It is validated for Flow Cytometry
Specifications
| SSX1 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| OTI1E10 | |
| Flow Cytometry | |
| Cancer/testis antigen 5.1, CT5.1cancer/testis antigen family 5, member 1, MGC150425, protein SSX1, sarcoma, synovial, X-chromosome-related 1, SSRC, synovial sarcoma, X breakpoint 1MGC5162 | |
| Full length human recombinant protein of human SSX1 (NP_005626) produced in E.coli. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 6756 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction