missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSX2IP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20488
This item is not returnable.
View return policy
Description
SSX2IP Polyclonal specifically detects SSX2IP in Mouse samples. It is validated for Western Blot.
Specifications
| SSX2IP | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal | |
| ADIP, afadin- and alpha-actinin-binding protein, Afadin DIL domain-interacting protein, FLJ10848, KIAA0923, MGC75026, SSX2-interacting protein, synovial sarcoma, X breakpoint 2 interacting protein | |
| Rabbit | |
| 71 kDa | |
| 0.1 mL | |
| Primary | |
| Mouse. Species reactivity based on sequence identity: Human (100%), Rhesus Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q9Y2D8 | |
| SSX2IP | |
| Recombinant protein encompassing a sequence within the center region of human SSX2IP. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 117178 | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction