missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSX2IP Antibody (OTI5C3), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74369AF405
This item is not returnable.
View return policy
Description
SSX2IP Monoclonal specifically detects SSX2IP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| SSX2IP | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SSX2IP | |
| Recombinant protein fragment corresponding to amino acids 1-272 of human SSX2IP (NP_054740) produced in E.coli. | |
| 0.1 mL | |
| 117178.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI5C3 | |
| Western Blot, Immunohistochemistry | |
| ADIP, afadin- and alpha-actinin-binding protein, Afadin DIL domain-interacting protein, FLJ10848, KIAA0923, MGC75026, SSX2-interacting protein, synovial sarcoma, X breakpoint 2 interacting protein | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction