missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSX5 Antibody (OTI3G6), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74372AF405
This item is not returnable.
View return policy
Description
SSX5 Monoclonal specifically detects SSX5 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| SSX5 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SSX5 | |
| Full length recombinant protein of human SSX5 (NP_066295) produced in HEK293T cells. | |
| 0.1 mL | |
| 6758.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI3G6 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| MGC9494, protein SSX5, synovial sarcoma, X breakpoint 5 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction