missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST2/IL-33R Antibody (2154B) [Janelia Fluor™ 525], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB5232JF525
This item is not returnable.
View return policy
Description
ST2/IL-33R Monoclonal antibody specifically detects ST2/IL-33R in Human samples. It is validated for Flow Cytometry,ELISA
Specifications
| ST2/IL-33R | |
| Monoclonal | |
| Janelia Fluor 525 | |
| DER4ST2growth stimulation-expressed, FIT-1, IL33R, interleukin 1 receptor-like 1, interleukin 1 receptor-related protein, interleukin-1 receptor-like 1, MGC32623, Protein ST2, ST2L, ST2V, T1homolog of mouse growth stimulation-expressed | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human ST-2/ IL-33R, Lys19-Phe328, Accession # Q01638 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, ELISA | |
| 2154B | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Angiogenesis | |
| 9173 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction