missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ST3GAL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553571
This item is not returnable.
View return policy
Description
Immunogen sequence: ATLPYLDSGA LDGTHRVKLV PGYAGLQRLS KERLSGKSCA CRRCMGDAGA SDWFDSHFDG NISPVWTREN MDLPPDVQRW WMMLQPQFKS HNTNEVLEKL FQIVPGENPY RFRDPHQCRR CAVVGNSGNL RGSGYGQDVD Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
ST3GAL2 is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The protein is normally found in the Golgi but can be proteolytically processed to a soluble form. This protein, which is a member of glycosyltransferase family 29, can use the same acceptor substrates as does sialyltransferase 4A. The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The encoded protein is normally found in the Golgi but can be proteolytically processed to a soluble form. This protein, which is a member of glycosyltransferase family 29, can use the same acceptor substrates as does sialyltransferase 4A.
Specifications
| ST3GAL2 | |
| Polyclonal | |
| Unconjugated | |
| ST3GAL2 | |
| 3-sialytransferase; AI429591; alpha 2,3-sialyltransferase ST3Gal II; alpha 2,3-ST 2; alpha-2,3-sialyltransferase; AW822065; beta-galactoside alpha-2,3-sialyltransferase 2; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 2; Gal-beta-1,3-GalNAc-alpha-2,3-sialyltransferase; gal-NAc6S; si:ch211-223o1.7; sialyltransferase 4B; sialyltransferase 4B (beta-galactosidase alpha-2,3-sialytransferase); sialyltransferase 4B (beta-galactoside alpha-2,3-sialyltransferase); sialyltransferase 5; sialyltransferase ST3Gal-II; Siat4b; SIAT4-B; siat5; ST3 beta-galactoside alpha-2,3-sialyltransferase 2; ST3Gal II; st3Gal II.2; St3gal2; st3gal2-r1; ST3GALA.2; ST3GalII | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6483 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16842 | |
| ST3GAL2 | |
| Recombinant protein corresponding to Human ST3GAL2. Recombinant protein control fragment (Product #RP-90346). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction