missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST3GAL4, Mouse anti-Human, Clone: 1F4, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00006484-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: FYFPIPEKKEPCLQGEAESKASKLFGNYSRDQPIFLRLEDYFWVKTPSAYELPYGTKGSEDLLLRVLAITSSSIPKNIQSLRCRRCVVVGNGHRLRNSSLSpecifications
| ST3GAL4 | |
| Monoclonal | |
| Unconjugated | |
| ST3 beta-galactoside alpha-2,3-sialyltransferase 4 | |
| CGS23/FLJ11867/FLJ46764/NANTA3/SAT3/SIAT4/SIAT4C/ST3GalIV/STZ | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1F4 | |
| In 1x PBS, pH 7.4 | |
| NM_006278 | |
| ST3GAL4 | |
| ST3GAL4 (NP_006269, 31 a.a. ∼ 130 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 6484 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction