missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 Antibody (001), Alexa Fluor™ 532 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89922AF532
This item is not returnable.
View return policy
Description
ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 Monoclonal antibody specifically detects ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 in Human samples. It is validated for ELISA, ELISA
Specifications
| ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| B-cell antigen CD75, beta-galactoside alpha-2,6-sialyltransferase 1, CMP-N-acetylneuraminate beta-galactosamide alpha-2,6-sialyltransferase, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 1, EC 2.4.99.1, MGC48859, Sialyltransferase 1, sialyltransferase 1 (beta-galactoside alpha-2,6-sialyltransferase), sialyltransferase 1 (beta-galactoside alpha-2,6-sialytransferase), SIAT1Alpha 2,6-ST 1, ST6 beta-galactosamide alpha-2,6-sialyltranferase 1, ST6Gal I, ST6GalI, ST6N | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 (P15907; Lys27-Cys406). | |
| 0.1 mL | |
| Golgi Apparatus Markers | |
| 6480 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 001 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction