Learn More
ST6GAL1 Mouse, anti-Human, Clone: 1F11G5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68633-1-IG-150UL
 View more versions of this product
Product Code. 18005575
  • € 410.00 / 150µL
Description
The ST6GAL1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ST6GAL1. This antibody recognizes human, pig antigen. The ST6GAL1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
ST6GAL1
1F11G5
Alpha 2,6 ST 1, B cell antigen CD75, CD75, Sialyltransferase 1, SIAT1, ST6Gal I, ST6GAL1, ST6GalI, ST6N
150 μL
6480
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human, Pig
150 kDa
IgG2b