Learn More
Invitrogen™ ST6GALNAC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139753
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: PQTKPSRHQR TENIKERSLQ SLAKPKSQAP TRARRTTIYA EPVPENNALN TQTQPKAHTT GDRGKEANQA PPEEQDKVPH TAQRAAWKSP EKEKTMVNTL SPRGQDAGMA SGRTEAQSWK SQDTKTTQGN Highest antigen sequence indentity to the following orthologs: Rat - 26%, Mouse - 25%.
Glycosylation of proteins affects cell-cell interaction, interactions with the matrix, and the functions of intracellular molecules. ST6GALNAC1 transfers a sialic acid, N-acetylneuraminic acid.
Specifications
| ST6GALNAC1 | |
| Polyclonal | |
| Unconjugated | |
| ST6GALNAC1 | |
| alpha-2,6-sialyltransferase ST6GalNAc I; alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 1; GalNAc alpha-2, 6-sialyltransferase I, long form; galNAc alpha-2,6-sialyltransferase I; HSY11339; sialyltransferase 7 ((alpha-N-acetylneuraminyl 2,3-beta-galactosyl-1,3)-N-acetyl galactosaminde alpha-2,6-sialyltransferase) A; sialyltransferase 7 ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase) A; sialyltransferase 7A; SIAT7A; SIAT7-A; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 1; ST6 GalNAc alpha-2,6-sialyltransferase 1; ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 1; ST6GalNAc I; ST6GALNAC1; ST6GALNAC1, alpha-2,6-sialyltransferase 1; ST6GalNAcI; STYI; UNQ543/PRO848 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55808 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NSC7 | |
| ST6GALNAC1 | |
| Recombinant protein corresponding to Human ST6GALNAC1. Recombinant protein control fragment (Product #RP-89970). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.