missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notch-3 Antibody (603532), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1559APCCY7
This item is not returnable.
View return policy
Description
Notch-3 Monoclonal antibody specifically detects Notch-3 in Human samples. It is validated for Flow Cytometry
Specifications
| Notch-3 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 603532 | |
| Flow Cytometry | |
| CADASIL, CASILneurogenic locus notch homolog protein 3, Notch (Drosophila) homolog 3, notch 3Notch homolog 3 (Drosophila), Notch homolog 3 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Notch-3, Ala40-Glu467, Accession # Q9UM47 | |
| 0.1 mL | |
| Cancer | |
| 4854 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction