missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin-1 Antibody (OTI8H2), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70172PCP
This item is not returnable.
View return policy
Description
Angiopoietin-1 Monoclonal antibody specifically detects Angiopoietin-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Angiopoietin-1 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI8H2 | |
| Western Blot, Immunohistochemistry | |
| AGP1, AGPT, Ang1, ANG-1, angiopoietin 1, angiopoietin-1, KIAA0003 | |
| Human recombinant protein fragment corresponding to amino acids 198-498 of human ANGPT1 (NP_001137) produced in E.coli. | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cell Cycle and Replication | |
| 284 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction